Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C1orf26 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524460

Catalog No. PIPA524460


Add to cart

Specifications

Specifications

C1orf26
Polyclonal
PBS with 0.09% sodium azide
Q5T5J6
C1orf26, HsSwt1, transcriptional protein SWT1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54823
Flow Cytometry, Western Blot
Unconjugated
SWT1
Liquid
SWT1
KLH conjugated synthetic peptide between 286-314 amino acids from the N-terminal region of human CA026
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit