Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase XI Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522339

Catalog No. PIPA522339


Add to cart

Description

Description

PA5-22339 targets CA11 in WB applications and shows reactivity with Human samples. The PA5-22339 immunogen is a recombit fragment corresponding to a region within amino acids 1 and 311 of CA11 (Uniprot IDNo.O75493). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Carbonic Anhydrase XI (CA11, CA XI) is a member of the carbonic anhydrases (CAs) family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. Carbonic Anhydrase XI participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. Further, Carbonic Anhydrase XI show extensive diversity in tissue distribution and in their subcellular localization. CA XI is likely a secreted protein, however, radical changes at active site residues completely conserved in CA isozymes with catalytic activity, make it unlikely that it has carbonic anhydrase activity. It shares properties in common with two other acatalytic CA isoforms, CA VIII and CA X. CA XI is most abundantly expressed in brain, and may play a general role in the central nervous system.
Specifications

Specifications

Carbonic Anhydrase XI
Polyclonal
Unconjugated
CA11
Liquid
CA11
Recombit fragment corresponding to a region within amino acids 1 and 311 of CA11 (Uniprot IDNo.O75493)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.81 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
O75493
UNQ211/PRO237, CARPX1, CA-RP II, CA-XI, CARP XI, CARP-2, carbonic anhydrase-related protein 11, carbonic anhydrase-related protein 2, carbonic anhydrase-related protein XI
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
770
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit