Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase XIV Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514190

Catalog No. PIPA514190

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CA14 belongs a large family of zinc metalloenzymes that catalyze the versible hydration of carbon dioxide. The protein is predicted to be a type I membrane protein and shares highest sequence similarity with the other transmembrane CA isoform, CA XII; however, they have different patterns of tissue-specific expression and thus may play different physiologic roles.
Specifications

Specifications

Carbonic Anhydrase XIV
Polyclonal
PBS with 0.09% sodium azide
Q9ULX7
Carbonic anhydrase 14; Carbonic anhydrase XIV; CA-XIV; Carbonate dehydratase XIV; UNQ690/PRO1335; CA14
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
23632
Flow Cytometry, Western Blot
Unconjugated
CA14
Liquid
CA14
KLH conjugated synthetic peptide between 51-80 amino acids from the N-terminal region of human CA14
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit