Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase VIII Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA542120
Description
Peptide sequence: TISQLQIEEF RRLRTHVKGA ELVEGCDGIL GDNFRPTQPL SDRVIRAAFQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%
Carbonic Anhydrase VIII Polyclonal antibody specifically detects Carbonic Anhydrase VIII in Human samples. It is validated for Western Blot.Specifications
Carbonic Anhydrase VIII | |
Polyclonal | |
Unconjugated | |
Ca8 | |
AW546993; Ca8; Cals; Cals1; CAMRQ3; Car8; carbonate dehydratase; carbonic anhydrase 8; carbonic anhydrase VIII; carbonic anhydrase-like sequence; carbonic anhydrase-like sequence 1; carbonic anhydrase-related protein; CA-related protein; CARP; CA-RP; CARP VIII; CA-RP VIII; CA-VIII; hypothetical protein LOC550233; LOW QUALITY PROTEIN: carbonic anhydrase-related protein; MGC120502; MGC99509; wdl; zgc:110118 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
767 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P35219 | |
Ca8 | |
Synthetic peptide directed towards the middle region of human CA8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction