Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaV2.2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521440

Catalog No. PIPA521440


Add to cart

Description

Description

PA5-21440 targets CACNA1B in ICC, IF, and WB applications and shows reactivity with Human samples. The PA5-21440 immunogen is a recombit fragment corresponding to a region within amino acids 2013 and 2209 of CACNA1B (Uniprot IDNo.Q00975). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

CaV2.2
Polyclonal
Unconjugated
CACNA1B
Liquid
CACNA1B
Recombit fragment corresponding to a region within amino acids 2013 and 2209 of CACNA1B (Uniprot IDNo.Q00975)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
0.43 mg/mL
PBS with 10% glycerol and 0.01% thimerosal; pH 7
Q00975
BIII, CACNL1A5, CACNN, Cav2.2, Cav2.2 voltage-gated Ca2+ channel, brain calcium channel III, calcium channel alpha12.2 subunit, calcium channel, L type, alpha-1 polypeptide, calcium channel, N type, calcium channel, voltage-dependent, L type, alpha 1B subunit, calcium channel, voltage-dependent, alpha 1B subunit, N type, voltage-dependent N-type calcium channel subunit alpha-1B, voltage-gated calcium channel alpha subunit Cav2.2, voltage-gated calcium channel subunit alpha Cav2.2
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
774
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit