Learn More
anti-CACNA2D1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP130556
Catalog No. NBP130556
Add to cart
Description
Description
CACNA2D1 Polyclonal antibody specifically detects CACNA2D1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
CACNA2D1
LYOPH
Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml
P54289
CACNA2, CACNL2Adihydropyridine-sensitive L-type, calcium channel alpha-2/delta subunit, calcium channel, L type, alpha 2 polypeptide, calcium channel, voltage-dependent, alpha 2/delta subunit 1, CCHL2A, MHS3, voltage-dependent calcium channel subunit alpha-2/delta-1, Voltage-gated calcium channel subunit alpha-2/delta-1
Rabbit
IgG
IgG purified
RUO
Primary
781
Reconstitute with 0.5 ml sterilized water.
Immunohistochemistry, Western Blot
Unconjugated
No buffer with No Preservative
Purified
CACNA2D1
A synthetic peptide from the n-terminal region of human CACNA2D1 (CACNL2A) conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is identical in rabbit and shares 90% identity with the corresponding sequences in mouse and rat
124.6 kDa
0.5 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Appears to be specific for CACNA2D1.
Human, Mouse