Learn More
anti-CACNB4, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP130728
Catalog No. NBP130728
Add to cart
Description
Description
CACNB4 Polyclonal antibody specifically detects CACNB4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
CACNB4
LYOPH
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
CACNB4
A synthetic peptide from the c-terminal region of human CACNB4 (CACB4, Voltage dependent L-type calcium channel subunit beta-4) conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in many species including mouse and rat.
IgG purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Appears to be specific for CACNB4.
Human, Mouse, Rat
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Sterilized water with No Preservative
calcium channel beta-4 subunit, calcium channel, voltage-dependent, beta 4 subunit, EJM, EJM4, EJM6, voltage-dependent L-type calcium channel subunit beta-4
Rabbit
IgG
0.5 mg
Neuroscience, Neurotransmission
Primary
785
Reconstitute with 0.5 ml sterilized water.