Learn More
anti-CACNG3, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP130696
Catalog No. NBP130696
Add to cart
Description
Description
CACNG3 Polyclonal antibody specifically detects CACNG3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
CACNG3
LYOPH
Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Purified
CACNG3
A synthetic peptide from the n-terminal region of human CACNG3 conjugated to an immunogenic carrier protein was used as the immunogen. The antigen is homologous in rat, mouse.
IgG purified
RUO
Primary
10368
Reconstitute with 0.5 ml sterilized water.
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Sterilized water with No Preservative
calcium channel, voltage-dependent, gamma subunit 3, Neuronal voltage-gated calcium channel gamma-3 subunit, voltage-dependent calcium channel gamma-3 subunit, voltage-gated calcium channel gamma subunit
Rabbit
IgG
0.5 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Appears to be specific for CACNG3.
Human, Mouse, Rat