Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcyclin Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522390

Catalog No. PIPA522390


Add to cart

Description

Description

PA5-22390 targets CacyBP in ICC, IF, and WB applications and shows reactivity with Human and mouse samples. The PA5-22390 immunogen is a recombit fragment corresponding to a region within amino acids 1 and 228 of CacyBP (Uniprot IDNo.Q9HB71). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

Calcyclin
Polyclonal
Unconjugated
CACYBP
Liquid
CACYBP
Recombit fragment corresponding to a region within amino acids 1 and 228 of CacyBP (Uniprot IDNo.Q9HB71)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Western Blot
0.94 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q9HB71, Q9CXW3
PNAS-107, GIG5, RP1-102G20.6, S100A6BP, SIP, S100A6-binding protein, Siah-interacting protein (SIP), calcyclin-binding protein, growth-inhibiting gene 5 protein, hCacyBP
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
12301, 27101
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit