Learn More
DFFB, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA130785
Description
Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain containing adapter molecules and members of the caspase family of proteases. These death signals finally cause the degradation of chromosomal DNA by activated DNase. A mouse DNase that causes DNA fragmentation was identified recently and designated CAD (for caspase activated deoxyribonuclease). The human homologue of mouse CAD was more recently identified by three groups independently and termed CPAN, DFF40, and human CAD, respectively. DFF45/ICAD is the inhibitory protein of DFF40/CAD and forms complex with DFF40/CAD. Upon cleavage of DFF45/ICAD by activated caspase, DFF40/CAD is released and activated and eventually causes the degradation of DNA in the nuclei. Activation of DFF40/CAD, which causes DNA degradation, is the hallmark of apoptotic cell death.
PA1-30785 detects CAD from mouse samples.PA1-30785 has been successfully used in Western blot applications.The PA1-30785 immunogen is: Synthetic peptide, corresponding to amino acids 314-329 of Mouse CAD.Specifications
DFFB/CAD | |
Polyclonal | |
Unconjugated | |
O54788 | |
DFFB | |
Synthetic peptide, corresponding to amino acids 314-329 of Mouse CAD. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.5mg/mL | |
Dffb | |
2410008J01Rik, AU018859, Cpad | |
Rabbit | |
IgG fraction | |
RUO | |
13368 | |
Store at 4°C | |
Purified |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.