Learn More
DFFB Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA130790
Catalog No. PA130790
Add to cart
Description
Description
PA1-30790 detects CAD from mouse, rat, human samples. PA1-30790 has been successfully used in Western blot applications. The PA1-30790 immunogen is: Peptide corresponding to amino acids 147 to 164 of mouseCAD (1). The sequence differs from human DFF40 by twoamino acids (3,4)
Apoptosis is related to many diseases and induced by a family of cell death receptors and their ligands. Cell death signals are transduced by death domain containing adapter molecules and members of the caspase family of proteases. These death signals finally cause the degradation of chromosomal DNA by activated DNase. A mouse DNase that causes DNA fragmentation was identified recently and designated CAD (for caspase activated deoxyribonuclease). The human homologue of mouse CAD was more recently identified by three groups independently and termed CPAN, DFF40, and human CAD, respectively. DFF45/ICAD is the inhibitory protein of DFF40/CAD and forms complex with DFF40/CAD. Upon cleavage of DFF45/ICAD by activated caspase, DFF40/CAD is released and activated and eventually causes the degradation of DNA in the nuclei. Activation of DFF40/CAD, which causes DNA degradation, is the hallmark of apoptotic cell death.
Specifications
Specifications
DFFB
Polyclonal
Unconjugated
DFFB
Liquid
Rabbit
IgG
100 μg
4° C
Primary
13368, 1677, 84359
Immunocytochemistry, Immunofluorescence, Western Blot
0.5 mg/mL
PBS with 0.02% sodium azide
O76075, O54788, Q99N34
DFFB
Peptide corresponding to amino acids 147 to 164 of murineCAD (1). The sequence differs from human DFF40 by twoamino acids (3,4)
IgG fraction
RUO
Antibody
Polyclonal
Human, Mouse, Rat