Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

calcitonin-related polypeptide alpha, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human CALCA protein.

Manufacturer:  Abnova Corporation H00000796B01

Catalog No. 89-005-020


Add to cart

Description

Description

Calcitonin is a peptide hormone synthesized by the parafollicular cells of the thyroid. It causes reduction in serum calcium, an effect opposite to that of parathyroid hormone (PTH; MIM 168450).[supplied by OMIM

Sequence: MGFQKFSPFLALSILVLLQAGSLHAAPFRSALESSPADPATLSEDEARLLLAALVQDYVQMKASELEQEQEREGSSLDSPRSKRCGNLSTCMLGTYTQDFNKFHTFPQTAIGVGAPGKKRDMSSDLERDHRPHVSMPQNAN
Specifications

Specifications

calcitonin-related polypeptide alpha
Unconjugated
No additive
NM_001741
CALC1|CGRP|CGRP-I|CGRP1|CT|KC|MGC126648
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
796
Western Blot
Mouse polyclonal antibody raised against a full-length human CALCA protein.
CALCA
Serum
CALCA
CALCA (NP_001732, 1 a.a. ~ 141 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit