Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcineurin Mouse, Unlabeled, Clone: 29, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610260

 View more versions of this product

Catalog No. BDB610260


Add to cart

Description

Description

Calcineurin is a Ca2+/calmodulin-dependent protein phosphatase. The active enzyme is a heterodimer of a large calmodulin-binding catalytic subunit A (61kDa) and a smaller Ca2+ binding subunit B (19kDa). Regions corresponding to the calmodulin-binding site, an autoinhibitory domain, and a putative subunit B binding site have been identified within the large subunit A. The activity of calcineurin is sensitive to immunosuppressants such as cyclosporin A (CsA) and tacrolimus (FK506). The study of FK506-mediated inhibition of nitric oxide formation has revealed that nitric oxide synthase (NOS) is a calcineurin substrate. Calcineurin dephosphorylates NOS and enhances its catalytic activity. Therefore, Calcineurin is an essential mediator for efficient T cell antigen receptor (TCR)-mediated T cell activation.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Calcineurin
29
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150μg
Cell Biology
Primary
Chicken, Frog, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.