Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcineurin A Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA515579

Catalog No. PIPA515579


Add to cart

Description

Description

This antibody is predicted to react with bovine and rat based on sequence homology.

PPP3CA is a calcium-dependent, calmodulin-stimulated protein phosphatase. This subunit may have a role in the calmodulin activation of calcineurin. PPP3CA dephosphorylates HSPB1 and SSH1.
Specifications

Specifications

Calcineurin A
Polyclonal
PBS with 0.09% sodium azide
Q08209, P63328
Serine/threonine protein phosphatase 2B catalytic subunit, alpha isoform; Calmodulin-dependent calcineurin A subunit, alpha isoform; CAM-PRP catalytic subunit; PPP3CA; CALNA, CNA
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
19055, 5530
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PPP3CA
Liquid
PPP3CA
KLH conjugated synthetic peptide between 21-51 amino acids from the N-terminal region of human Calcineurin (PPP3CA)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit