Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RCAN1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527132

Catalog No. PIPA527132


Add to cart

Description

Description

PA5-27132 targets Calcipressin 1 in WB applications and shows reactivity with Human samples. The PA5-27132 immunogen is a synthetic peptide corresponding to a region within amino acids 164 and 252 of Calcipressin 1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene interacts with calcineurin A and inhibits calcineurin-dependent signaling pathways, possibly affecting central nervous system development. This gene is located in the minimal candidate region for the Down syndrome phenotype, and is overexpressed in the brain of Down syndrome fetuses. Chronic overexpression of this gene may lead to neurofibrillary tangles such as those associated with Alzheimer disease. Three transcript variants encoding three different isoforms have been found for this gene.
Specifications

Specifications

RCAN1
Polyclonal
Unconjugated
RCAN1
Liquid
RCAN1
Synthetic peptide corresponding to a region within amino acids 164 and 252 of Calcipressin 1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
PBS with 20% glycerol and 0.01% thimerosal; pH 7
P53805
ADAPT78, CSP1, DSC1, DSCR1, MCIP1, RCN1, Down syndrome candidate region 1, Down syndrome critical region gene 1, calcipressin-1, calcium and oxidant-inducible mRNA, down syndrome critical region protein 1, modulatory calcineurin-interacting protein 1, myocyte-enriched calcineurin-interacting protein 1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1827
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit