Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CACNB3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112670

Catalog No. 01-674-824


Add to cart

Description

Description

A suggested positive control for this product is rat brain lysate.

The expression of multiple classes of voltage dependent calcium channels (VDCC’s) allows neurons to tailor calcium signaling to functionally discrete cellular regions. Although N-Type VDCC’s exist before birth which is consistent with a role in migration, most N-Type VDCC’s subunit expression is postnatal and is important in the genesis of synaptic transmission in discrete hippocampal fields.
Specifications

Specifications

CACNB3
Polyclonal
PBS with 0.08% sodium azide
P54284
CACNLB3
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
784
Western Blot
Unconjugated
CACNB3
Liquid
CACNB3
Synthetic peptide, conjugated to KLH, corresponding to the rat beta3 calcium channel.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit