Learn More
anti-Calcium Channel, Voltage Gated 1C ; Polyclonal; Millipore™ (Chemicon™)
Manufacturer:  Millipore Sigma AB5156/DEL
 View more versions of this product
Catalog No. 50-173-688
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Calcium Channel, Voltage Gated 1C
Affinity Purified
Purified peptide (C)TTKI NMDDL QPSEN EDKS (CNC1) from 848-865 of α 1C subunit of rat brain voltage-gated Dlcium channel. Epitope loDtion is in the intracellular loop between domains II and III.
RUO
Primary
Human,Murine,Rabbit,Rat
Unlabeled
Rabbit
200μL
Neuroscience
Polyclonal