Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CALCOCO2 Mouse anti-Human, Clone: P17410, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA518290

Catalog No. PIMA518290


Add to cart

Description

Description

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The protein encoded by this gene is a subunit of nuclear domain 10 (ND10) bodies. ND10 bodies are nuclear domains appearing immunohistochemically as ten dots per nucleus. They are believed to be associated with the nuclear matrix on the basis of their resistance to nuclease digestion and salt extraction. ND10 proteins are removed from the nucleus by herpes simplex virus-1 infection and may have a role in viral life cycles.
Specifications

Specifications

CALCOCO2
Monoclonal
1 mg/mL
PBS with no preservative
Q13137
NDP52, calcium binding and coiled-coil domain 2
Mouse
IgG2b
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
10241
Western Blot
P17410
Unconjugated
CALCOCO2
Liquid
CALCOCO2
Recombit fragment corresponding to a region within amino acids 174 and 430 of NDP52
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit