Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CALML5, Mouse, Clone: 2F10, Abnova™

Mouse monoclonal antibody raised against a full-length recombinant CALML5.

Manufacturer:  Abnova Corporation H00051806M16

Catalog No. 89-002-633


Add to cart

Description

Description

This gene encodes a novel calcium binding protein expressed in the epidermis and related to the calmodulin family of calcium binding proteins. Functional studies with recombinant protein demonstrate it does bind calcium and undergoes a conformational change when it does so. Abundant expression is detected only in reconstructed epidermis and is restricted to differentiating keratinocytes. In addition, it can associate with transglutaminase 3, shown to be a key enzyme in the terminal differentiation of keratinocytes. [provided by RefSeq

Sequence: MAGELTPEEEAQYKKAFSAVDTDGNGTINAQELGAALKATGKNLSEAQLRKLISEVDGDGDGEISFQEFLTAARKARAGLEDLQVAFRAFDQDGDGHITVDELRRAMAGLGQPLPQEELDAMIREADVDQDGRVNYEEFARMLAQE
Specifications

Specifications

CALML5
2F10
Mouse monoclonal antibody raised against a full-length recombinant CALML5.
CALML5
Liquid
CALML5
CALML5 (AAH39172, 1 a.a. ~ 146 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 ug
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
In 1X PBS, pH 7.4
BC039172
CLSP
Mouse
IgG2a Kappa
RUO
Primary
51806
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit