Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calmodulin Binding Protein Epitope Tag Rabbit anti-Vertebrates, Clone: C16T, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 04932
Description
This Anti-Calmodulin Binding Protein Epitope Tag Antibody, clone C16T is validated for use in western blotting for the detection of Calmodulin Binding Protein Epitope Tag.Specifications
Calmodulin Binding Protein Epitope Tag | |
Monoclonal | |
Unconjugated | |
Rabbit | |
100 μL | |
CALM, CALML2, CAM, CAM1, CAM2, CAM3, CAMB, CAMC, CAMI, CAMIII, CaM, DD132, PHKD | |
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles. | |
IgG |
Western Blot | |
C16T | |
Cultured supernatant with 0.05% sodium azide. | |
KLH-conjugated, synthetic peptide corresponding to the Calmodulin Binding Protein (CBP) epitope tag found in several E. coli expression vectors. | |
Primary | |
Vertebrates | |
Supernatant |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only