Learn More
Calmodulin Binding Protein Epitope Tag Rabbit anti-Vertebrates, Clone: C16T, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma 05932
Catalog No. 05-932-MI
Add to cart
Description
Description
Anti-Calmodulin Binding Protein Epitope Tag Antibody, clone C16T is an antibody against Calmodulin Binding Protein Epitope Tag for use in western blotting.
Specifications
Specifications
Calmodulin Binding Protein Epitope Tag
C16T
Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide as a preservative.
Rabbit
IgG
100μL
Primary
Vertebrate
Western Blot
Unconjugated
Purified
KLH-conjugated, synthetic peptide corresponding to the Calmodulin Binding Protein (CBP) epitope tag found in several E. coli expression vectors
Protein A purfied
−20°C for 24 months. Aliquot to avoid freeze/thaw cycles.
Monoclonal