Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calmodulin Mouse anti-Bovine, Human, Mouse, Pig, Rat, Vertebrates, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05173
Description
Detect Calmodulin using this Anti-Calmodulin Antibody validated for use in western blotting and immunoprecipitation.Specifications
Calmodulin | |
Monoclonal | |
Purified mouse IgG1 in storage buffer containing 0.1MTris-glycine, pH 7.4, 0.15MNaCl, 0.05% sodium azide. Frozen solution. | |
CALM1, CALML2, CALML2, CALM3, CALM, CaM, CALM2, CAM2, CAMC, CAM3, DD132, CAMIII, PHKD, CAMI, CAMB, CAM, CAM1 | |
Synthetic peptide corresponding to the 21 carboxy-terminal amino acids (128-148) of bovine calmodulin, conjugated with tyroglobulin. | |
100 μg | |
Bovine, Human, Mouse, Porcine, Rat, Vertebrates | |
Purified |
Immunoprecipitation, Western Blot | |
Unconjugated | |
NP_008819 | |
Mouse | |
Protein G chromatography | |
Primary | |
Stable for 1 year at from date of receipt. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only