Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpain S1/S2 Mouse anti-Human, Agarose, Clone: 28F3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA112432

Catalog No. 01-674-302


Add to cart

Description

Description

This antibody specifically recognizes the 28 to 30 kD small subunit of Calpain 1 and Calpain 2 (mu calpain and m calpain).

The calpains are calcium-dependent cysteine proteases that are widely expressed in mammalian systems. Both m-calpain (calpain II) and m-calpain (calpain I) are composed of an 80 kD subunit and a 30 kD subunit. Whereas the 30 kDa subunit is shared by both enzymes, the larger catalytic subunits are different and exhibit the distinct Ca++ requirements that are suggested by their names. Whereas m-calpain requires millimolar (mM) levels of calcium, m-calpain is active at micromolar (mM) concentrations of Ca++. In addition to the ubiquitously expressed m- and m- calpains, some tissue-specific calpains have been identified. The calpains appear to serve multiple physiological roles, and ideas concerning the functions of these enzymes are in a state of rapid flux.
Specifications

Specifications

Calpain S1/S2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 3mM sodium azide; pH 7.5
P04632, Q96L46
CAPNS1, CAPNS2
Human Calpain protein purified from placenta.
Protein A/G
RUO
Antibody
Monoclonal
Human
Immunoprecipitation
28F3
Agarose
CAPNS2
Slurry
Mouse
IgG1
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
826, 84290
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit