Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calpain 2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527720

Catalog No. PIPA527720


Add to cart

Description

Description

PA5-27720 targets Calpain 2 in IF, IHC (P), and WB applications and shows reactivity with Human and mouse samples. The PA5-27720 immunogen is recombit fragment corresponding to a region within amino acids 162 and 383 of Human Calpain 2. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The calpains, calcium-activated neutral proteases, are nonlysosomal, intracellular cysteine proteases. The mammalian calpains include ubiquitous, stomach-specific, and muscle-specific proteins. The ubiquitous enzymes consist of heterodimers with distinct large, catalytic subunits associated with a common small, regulatory subunit. This gene encodes the large subunit of the ubiquitous enzyme, calpain 2. Multiple heterogeneous transcriptional start sites in the 5 ft. UTR have been reported. Two transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

Calpain 2
Polyclonal
Unconjugated
CAPN2
Liquid
CAPN2
Recombit fragment corresponding to a region within amino acids 162 and 383 of Human Calpain 2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
P17655, O08529
CANP2, CANPL2, CANPml, mCANP, CANP 2, M-calpain, calcium-activated neutral proteinase 2, calpain 2, large [catalytic] subunit, calpain 2, large subunit, calpain M-type, calpain large polypeptide L2, calpain, large polypeptide L2, calpain-2 catalytic subunit, calpain-2 large subunit, millimolar-
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
12334, 824
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit