Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calpain 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP131756
Description
Calpain 3 Polyclonal specifically detects Calpain 3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Calpain 3 | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
CAPN3 | |
Recombinant protein encompassing a sequence within the center region of human Calpain 3. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
825 | |
Human | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
0.43mg/mL | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
calpain 3, (p94), Calpain L3, Calpain p94, calpain, large polypeptide L3, calpain-3, CANP 3, CANP3MGC4403, CANPL3, EC 3.4.22, EC 3.4.22.54, LGMD2, LGMD2A, MGC10767, MGC11121, MGC14344, Muscle-specific calcium-activated neutral protease 3, muscle-specific calcium-activated neutral protease 3 large subunit, NCL1, nCL-1large [catalytic] subunit, New calpain 1 | |
Rabbit | |
94 kDa | |
0.1 mL | |
Primary | |
Tested in human. Not yet tested in any other species. Expected to cross react based on sequence identity:Mouse (86%), Pig (88%), Rat (89%), Bovine (88%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction