Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Calreticulin Ab-2; Polyclonal; ™ (Upstate™)
Supplier: MilliporeSigma™ 06661/DEL

Description
Waser et al., J. Cell Biol. 138:547, 1997
Immunocytochemistry, Immunohistochemistry, Western Blotting
Specifications
Calreticulin Ab-2 | |
Unconjugated | |
Peptide (QAKDEL) corresponding to the C-terminus (amino acids 412-417) of human calreticulin | |
RUO | |
Primary | |
Semi-purified |
Polyclonal | |
Rabbit | |
200 μg | |
Cell Signaling, Endocrine/Metabolism | |
Bovine, Human, Mouse, Porcine, Rat, Xenopus | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title