Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-Calreticulin Clone: FMC75,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ 20891250UG
Description
Specifically detects Calreticulin Clone: FMC75 in Human, Monkey samples, and it is validated for Enzyme Immunoassay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immuno Blotting
Calreticulin is a calcium binding protein found primarily in the ER and the sarcoplasmic reticulum, although it also has been detected in the nucleus and nuclear envelope. It has the conserved KDEL sequence at its C-terminus for retention in the ER. It contains both high and low affinity calcium binding sites, is involved in calcium storage and regulation. The calcium binding at the C-terminus of calreticulin has been postulated to be involved with regulating the interaction of KDEL proteins with the KDEL receptor. Calreticulin also acts as a molecular chaperone for protein folding, in concert with Grp78 and Grp94.Specifications
Calreticulin | |
Monoclonal | |
Unconjugated | |
Mouse | |
50 μg | |
Primary | |
Human, Monkey | |
Purified |
ELISA, Flow Cytometry, Immunoblot, Immunoblot, Immunocytochemistry, Immunohistochemistry | |
FMC75 | |
In PBS, 0.1mM PMSF, 50% glycerol, pH 7.2. | |
a recombinant protein consisting of human calreticulin fused to maltose binding protein | |
RUO | |
Recognizes the ∽63kDa (apparent MW) calreticulin protein in Molt 4, MCF-7, HeLa, K562, A431, Jurkat, Hs67, Hs198, and vero cells. | |
−20°C | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only