Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calreticulin Goat anti-Human, Mouse, Rabbit, Rat, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA184481

Catalog No. PA184481


Add to cart

Description

Description

Goat anti calreticulin antibodyrecognizes calreticulin, a 418 aminoacid approximately 60 kDa Ca2+ binding chaperone involved in protein modification and folding within the endoplasmic reticulum (UniProt: P15253).

Calr or Calreticulin is a calcium-binding chaperone that promotes folding, oligomeric assembly and quality control in the endoplasmic reticulum (ER) via the calreticulin/calnexin cycle. Calr, a lectin, interacts transiently with almost all of the monoglucosylated glycoproteins that are synthesized in the ER. Calr also interacts with with the DNA-binding domain of NR3C1 and mediates its nuclear export. It may also be involved in maternal gene expression regulation and oocyte maturation via the regulation of calcium homeostasis.
Specifications

Specifications

Calreticulin
Polyclonal
whole serum diluted in glycine buffered saline with 0.09% sodium azide
P27797, P14211, P15253, P18418
CALR
Purified rabbit skeletal muscle calreticulin.
100 μL
4°C or -20°C if preferred
Primary
100009050, 12317, 64202, 811
Immunohistochemistry (Frozen), Western Blot
Unconjugated
CALR
Liquid
Goat
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Rabbit, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit