Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calretinin Mouse, Unlabeled, Clone: 34, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610908

Catalog No. BDB610908


Add to cart

Description

Description

A low level of intracellular free Ca2+ is essential for a variety of cellular functions and is mediated by the sequestration of Ca2+ in the ER, by the action of plasma membrane Ca2+ pumps, and by Ca2+ binding proteins. The Ca2+ binding protein, Calretinin, is a member of the calmodulin superfamily whose members promote calcium homeostasis by acting as buffers of intracellular free Ca2+. Although Calretinin is expressed in certain populations of neurons throughout the central and peripheral nervous systems, it is primarily expressed in the olfactory bulb and auditory pathways. Calretinin most closely resembles calbindin D28k, another member of the calmodulin family. These two proteins share 58% amino acid identity and contain six EF-hand Ca2+ binding motifs. Excess Calretinin or calbindin results in decreased transcription from PCE1, a calcium responsive element in the calmodulin II promoter. This supports the role of Calretinin as a regulator of intracellular free Ca2+ levels. Furthermore, truncated forms of Calretinin have been found in certain tumor cells, but are absent from normal cells which express wild type Calretinin. Thus, these isoforms may have a role in tumorigenesis.

Specifications

Specifications

Calretinin
34
Unconjugated
Affinity Purified
Rat Calretinin aa. 38-151
50μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit