Learn More
Calretinin Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA137222
Description
This antibody is predicted to react with mouse based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is mesothelioma.
Calretinin is an intracellular calcium-binding protein belonging to the troponin C superfamily characterized by a structural mitif described as the EF-hand domain. The immunohistochemical detection of calretinin in developing cerebellum is restricted to the later stages indicated by weak staining from week 21 of gestation, in Purkinje and basket cells and in neurons of the dentate nucleus. The intensity of staining increases as the cerebellum matures. In tumors, calretinin has been detected in mesotheliomas and some pulmonary adenocarcinomas.
Specifications
Calretinin | |
Polyclonal | |
CALB2 | |
CAL2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
794 | |
4° C, do not freeze | |
Liquid |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
P22676 | |
CALB2 | |
Recombit full length mouse calretinin protein | |
7 mL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.