Learn More
Calsenilin/DREAM Mouse anti-Human, Mouse, Rat, Clone: 40A5, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05756
Catalog No. 05-756-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect Calsenilin/DREAM using this Anti-Calsenilin/DREAM Antibody, clone 40A5 validated for use in immunoprecipitation and western blotting.
Specifications
Specifications
Calsenilin/DREAM
40A5
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Purified
Murine
Ascites
−20°C for 24 months.
Monoclonal
Immunoprecipitation, Western Blot
Unconjugated
NM_013434
KCNIP3, MGC18289, KChIP3, DREAM, CSEN, calsenilin, Calsenilin, KCHIP3
Full length GST fusion protein of human Calsenilin/DREAM
100μL
Primary
Human, Murine, Rat