Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMK1D Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA141243

Catalog No. 01-675-597


Add to cart

Description

Description

Suggested positive control: antigen standard for CAMK1D (transient overexpression lysate).

Calcium/clamodulin-dependent protein kinase 1D-beta isoform is a novel calmodulin-kinase that exhibits Ca (2+)/CaM-dependent activity. CAM-Kkalpha upregulates CAMK1D-b activity through phosphorylation of its Thr180.
Specifications

Specifications

CAMK1D
Polyclonal
Unconjugated
CAMK1D
Liquid
CAMK1D
Synthetic peptide corresponding to residues 10-50 of human CaMK 1D-beta.
Protein G
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.2% gelatin and 0.05% sodium azide
Q8IU85
CAMKI, MGC120317, MGC120318
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
57118
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit