Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CaM Kinase II, Polyclonal, ™ (Upstate™) Non-distribution product as customer accommodation.

Manufacturer:  MilliporeSigma™ 071496/DEL

Catalog No. 50-172-281

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

The Ca2+/Calmodulin-Dependent Kinase family includes CaM Kinases I through IV, Phosphorylase Kinase, and MLCK. The CaM Kinases are inactive in the absence of stimuli; binding to Ca2+/Calmodulin induces conformational changes and activation. CaM Kinases have broad substrate specificity, with many cellular functions.

Host Species:Rabbit
Clone:
Isotype:IgG
Species Reactivity:Human, Rat, Mouse, Bovine, Hamster, Guinea Pig, Chicken Rabbit
Epitope:N-terminus
Immunogen:Synthetic peptide corresponding to the N-terminus of Rat CaMK II
Molecular Weight:50-72kDa, corresponding to apparent molecular mass of CaMKII isoforms.

Western Blotting

Specifications

Specifications

CaM Kinase II
Affinity Purified
IgG
RUO
Primary
Bovine, Chicken, Guinea Pig, Hamster, Human, Murine, Rabbit, Rat
Unconjugated
Rabbit
100μL
Cell Signaling
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit