Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaM Kinase IV Mouse, Unlabeled, Clone: 26, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610275

 View more versions of this product

Catalog No. BDB610275


Add to cart

Description

Description

CaM-kinase IV (Ca2+calmodulin-dependent protein kinase IV, also described as CaM-kinase Gr) is activated through the binding of Ca2+/CaM and by phosphorylation. This kinase has high sequence homologies with the catalytic and regulatory domains of CaM-kinase II. CaM-kinase IV is a monomer which is highly expressed in cerebellum, thymus, and testis. Its subcellular distrubution includes localization in both the synaptic regions of the molecular layer of the cerebellum, and the nuclei of cerebellar granule cells. CaM-kinase IV has an autoinhibitory domain within residues 305-321 that can suppress kinase activity in the absence of Ca2+/CaM. This type of domain is common in many other CaM-dependent kinases and phosphatases. CaM-kinase IV does not appear to be significantly activated by autophosphorylation but it can be activated approximately 5-10-fold when phosphorylated by CaM-kinase IV kinase.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

CaM Kinase IV
26
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG1
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.