Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaM Kinase VI (CPG16; Candidate Plasticity-related Gene 16) Mouse, Unlabeled, Clone: 5, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 611736

 View more versions of this product

Catalog No. BDB611736


Add to cart

Description

Description

Central nervous system plasticity is essential to the processes of learning and memory. Plasticity involves changes in synaptic transmission efficiency and results from the activation of excitatory glutamate receptors. Analysis of changes in gene expression during kainate-induced hippocampal plasticity led to the identification of a Candidate Plasticity-related Gene (CPG16). CPG16 protein is a Ser/Thr protein kinase that is also closely related to the Ca2+/calmodulin- dependent protein kinases (CaM kinases). CPG16 is 98% identical to the C-terminal region of the product of the KIAA0369 gene, a 96kDa protein whose N-terminal region strongly resembles the neural protein, doublecortin. In resting cells, Cam Kinase VI (CPG16) is found in the cytoplasm and has relatively low level of phosphorylation activity. However, treatment of cells with cAMP-elevating agents (forskolin, 8-bromo-cAMP) significantly enhances this phosphorylation activity and induces a partial shift of Cam Kinase VI (CPG16) into the nucleus. In addition, overexpression of Cam Kinase VI (CPG16) inhibits cAMP-stimulated CREB activity. Thus, Cam Kinase VI (CPG16) is Ser/Thr protein kinase that is thought to be a downstream negative regulator of cAMP/PKA-induced transcription.

Host Species: Mouse
Clone: 5
Isotype: IgG1
Species Reactivity [for Features Main]: Rat
Immunogen: Rat CPG16 aa. 330-424

Immunofluorescence, Western Blotting

Specifications

Specifications

CaM Kinase VI (CPG16; Candidate Plasticity-related Gene 16)
5
Unconjugated
Murine
IgG1
RUO
Primary
Murine, Rat
Immunofluorescence, Western Blot
250μg/mL
Affinity Purified
Rat CPG16 aa. 330-424
50μg
Cell Biology
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.