Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaMKI Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528355

Catalog No. PIPA528355


Add to cart

Description

Description

PA5-28355 targets CaMK1 in IF and WB applications and shows reactivity with Human samples. The PA5-28355 immunogen is recombit fragment corresponding to a region within amino acids 99 and 312 of Human CaMK1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Calcium/calmodulin-dependent protein kinase I is expressed in many tissues and is a component of a calmodulin-dependent protein kinase cascade. Calcium/calmodulin directly activates calcium/calmodulin-dependent protein kinase I by binding to the enzyme and indirectly promotes the phosphorylation and synergistic activation of the enzyme by calcium/calmodulin-dependent protein kinase I kinase.
Specifications

Specifications

CaMKI
Polyclonal
Unconjugated
CAMK1
Liquid
CAMK1
Recombit fragment corresponding to a region within amino acids 99 and 312 of Human CaMK1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.78 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q14012, Q91YS8
CAMKI, caM kinase I alpha, caM-KI, caMKI-alpha, calcium/calmodulin-dependent protein kinase type 1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
52163, 8536
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit