Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMK1D Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514030

Catalog No. PIPA514030

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CAMK1-like protein kinase belongs to a proposed calcium-triggered signaling cascade. This protein is expressed in polymorphonuclear leukocytes (PMNs) and may be part of the chemokine signal transduction pathway that regulates granulocyte function. CAMK1-like protein kinase may play a role in apoptosis of erythroleukemia cells. It activates MAP kinase MAPK3, and in vitro, phosphorylates transcription factor CREM isoform Beta and probably CREB1.
Specifications

Specifications

CAMK1D
Polyclonal
PBS with 0.09% sodium azide
Q8IU85
Calcium/calmodulin-dependent protein kinase type 1D; CaM kinase ID; CaM kinase I delta; CaMKI-delta; CaM-KI delta; CaMKI delta; Camk1D; CamKI-like protein kinase; CKLiK; CamKI-like protein kinase; CAMK1D
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57118
Immunohistochemistry, Western Blot
Unconjugated
CAMK1D
Liquid
CAMK1D
KLH conjugated synthetic peptide between 302-333 amino acids from the C-terminal region of human CAMK1D (CAMK1 delta)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit