Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

calcium/calmodulin-dependent protein kinase II gamma, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human CAMK2G protein.

Manufacturer:  Abnova Corporation H00000818B01

Catalog No. 89-005-028


Add to cart

Description

Description

The product of this gene belongs to the Serine/Threonine protein kinase family, and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. Calcium signaling is crucial for several aspects of plasticity at glutamatergic synapses. In mammalian cells the enzyme is composed of four different chains: alpha, beta, gamma, and delta. The product of this gene is a gamma chain. Six alternatively spliced variants that encode six different isoforms have been characterized to date. Additional alternative splice variants that encode different isoforms have been described, but their full-length nature has not been determined. [provided by RefSeq

Sequence: MATTATCTRFTDDYQLFEELGKGAFSGVRRCVKKTPTQEYAAKIINTKKLSARDHQKLEREARICRLLKHPNIVRLHDSISEEGFHYLVFDLVTGGELFEDIVAREYYSEADASHCIHQILESVNHIHQHDIVHRDLKPENLLLASKCKGAAVKLADFGLAIEVQGEQQAWFGFAGTPGYLSPEVLRKDPYGKPVDIWACGVILYILLVGYPPFWDEDQHKLYQQIKAGAYDFPSPEWDTVTPEAKNLINQMLTINPAKRITADQALKHPWVCQRSTVASMMHRQETVECLRKFNARRKLKGAILTTMLVSRNFSVGRQSSAPASPAASAAGLAGQAAKSLLNKKSDGGVKKRKSSSSVHLMEPQTTVVHNATDGIKGSTESCNTTTEDEDLKVRKQEIIKITEQLIEAINNGDFEAYTKICDPGLTSFEPEALGNLVEGMDFHKFYFENLLSKNSKPIHTTILNPHVHVIGEDAACIAYIRLTQYIDGQGRPRTSQSEETRVWHRRDGKWLNVHYHCSGAPAAPLQ
Specifications

Specifications

calcium/calmodulin-dependent protein kinase II gamma
Unconjugated
No additive
BC034044
CAMK|CAMK-II|CAMKG|FLJ16043|MGC26678
Mouse
50 uL
Kinases
Y
Polyclonal
Human, Rat
Immunofluorescence, Western Blot
Mouse polyclonal antibody raised against a full-length human CAMK2G protein.
CAMK2G
Serum
CAMK2G
CAMK2G (AAH34044.1, 1 a.a. ~ 527 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
818
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit