Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaMKII gamma Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514035

Catalog No. PIPA514035


Add to cart

Description

Description

CaM-kinase II (CAMK2) is a prominent Ser/Thr protein kinase in the central nervous system that may function in long-term potentiation and neurotransmitter release. Likely autophosphorylation of Thr-286 allows the kinase to switch from a calmodulin-dependent to a calmodulin-independent state. CAMK2 is composed of four different chains: alpha, beta, gamma, and delta. The different isoforms assemble into homo- or heteromultimeric holoenzymes composed of 8 to 12 subunits.
Specifications

Specifications

CaMKII gamma
Polyclonal
PBS with 0.09% sodium azide
Q13555, Q923T9
Calcium/calmodulin-dependent protein kinase type II gamma chain; CaM-kinase II gamma chain; CaM kinase II gamma subunit; CaMK-II gamma subunit; CAMK2G
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
12325, 818
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CAMK2G
Liquid
CAMK2G
KLH conjugated synthetic peptide between 309-338 amino acids from the C-terminal region of human CAMK2G (CAMK2 gamma)
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit