Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CaMKIIN2 (p Thr286), Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP150259

Catalog No. NBP150259


Add to cart

Description

Description

CaMKIIN2 p Thr286 Polyclonal antibody specifically detects CaMKIIN2 p Thr286 in Human, Mouse, Rat, Xenopus samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications

Specifications

CaMKIIN2 (p Thr286)
Unconjugated
10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with 50% glycerol
calcium/calmodulin-dependent protein kinase II inhibitor 2, CaM-KII inhibitory protein, CaM-KIIN, CAMKIIN
Rabbit
IgG
0.05 ml
Phospho Specific
Primary
94032
Human, Mouse, Rat, Xenopus
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200
Affinity Purified
CAMK2N2
Phosphopeptide corresponding to amino acid residues surrounding the phosphoThr286 found in rat brain CaM Kinase II.
Immunogen affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Specific for the ~50k a-CaM Kinase II and the ~60k R-CaM Kinase II proteins phosphorylated at Thr286. Immunolabeling is blocked by the X-phosphatase treatment
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only