Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CaMKIV Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528346

Catalog No. PIPA528346


Add to cart

Description

Description

PA5-28346 targets CaMKIV in IHC (P) and WB applications and shows reactivity with Human samples. The PA5-28346 immunogen is recombit fragment corresponding to a region within amino acids 65 and 473 of Human CaMKIV. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The product of this gene belongs to the serine/threonine protein kinase family, and to the Ca(2+)/calmodulin-dependent protein kinase subfamily. This enzyme is a multifunctional serine/threonine protein kinase with limited tissue distribution, that has been implicated in transcriptional regulation in lymphocytes, neurons and male germ cells.
Specifications

Specifications

CaMKIV
Polyclonal
Unconjugated
CAMK4
Liquid
CAMK4
Recombit fragment corresponding to a region within amino acids 65 and 473 of Human CaMKIV
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.92 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q16566
CaMK IV, CaMK-GR, IV, caMK, CAM kinase IV, CAM kinase- GR, brain Ca(2+)-calmodulin-dependent protein kinase type IV, brain Ca++-calmodulin-dependent protein kinase type IV, caM kinase-GR, calcium/calmodulin-dependent protein kinase type IV, calcium/calmodulin-dependent protein kinase type
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
814
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit