Learn More
CAMLG Mouse anti-Human, Clone: 3F12, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA518533
Catalog No. PIMA518533
Add to cart
Description
Description
Peptide Sequence: MESMAVATDG GERPGVPAGS GLSASQRRAE LRRRKLLMNS EQRINRIMGF HRPGSGAEEE SQTKSKQQDS DKLNSLSVPS VSKRVVLGDS VSTGTTDQQG
The immunosuppressant drug cyclosporin A blocks a calcium-dependent signal from the T-cell receptor (TCR) that normally leads to T-cell activation. When bound to cyclophilin B, cyclosporin A binds and inactivates the key signaling intermediate calcineurin. The protein encoded by this gene functions similarly to cyclosporin A, binding to cyclophilin B and acting downstream of the TCR and upstream of calcineurin by causing an influx of calcium. This integral membrane protein appears to be a new participant in the calcium signal transduction pathway, implicating cyclophilin B in calcium signaling, even in the absence of cyclosporin.
Specifications
Specifications
CAMLG
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
P49069
CAML, calcium signal-modulating cyclophilin ligand, calcium-modulating cyclophilin ligand, calcium-signal modulating cyclophilin ligand, cyclophilin B-binding protein
Mouse
IgG2b, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
819
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
3F12
Unconjugated
CAMLG
Liquid
CAMLG
CAMLG (NP_001736, 1 a.a. ∽ 100 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human