Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAMLG Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525779

Catalog No. PIPA525779

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CAMLG
Polyclonal
PBS with 0.09% sodium azide
P49069
CAML, calcium signal-modulating cyclophilin ligand, calcium-modulating cyclophilin ligand, calcium-signal modulating cyclophilin ligand, cyclophilin B-binding protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
819
Flow Cytometry, Western Blot
Unconjugated
CAMLG
Liquid
CAMLG
KLH conjugated synthetic peptide between 111-138 amino acids from the central region of human CAMLG
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit