Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CAMP/LL37/FALL39/Cathelicidin, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP176864

Catalog No. NBP176864


Add to cart

Description

Description

CAMP/LL37/FALL39/Cathelicidin Polyclonal antibody specifically detects CAMP/LL37/FALL39/Cathelicidin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen (Negative).
Specifications

Specifications

CAMP/LL37/FALL39/Cathelicidin
1 mg/mL
Western Blot 1-2 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 20 ug/mL, Immunohistochemistry-Paraffin 5 ug/ml
P49913
18 kDa cationic antimicrobial protein, CAP18CRAMP, cathelicidin antimicrobial peptide, FALL-39, FALL39CAP-18, hCAP-18, LL37
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
820
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PBS with 0.02% Sodium Azide
Affinity Purified
CAMP
Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of the human Cathelicidin. The immunogen is located within amino acids 50 - 100 of Cathelicidin.
19 kDa
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only