Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-CAMPY SP AB FITC  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 53300060
Affinity purified polyclonal antibody to the Campylobacter species made in Goat and labeled with fluorescein isothiocyanate (FITC). Isolated from a serum pool of goats immunized with different strains of heat-killed whole cells of Camplyobacter.
Catalog No. 50-669-19