Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Campylobacter jejuni, Polyclonal, Invitrogen™

Catalog No. PIPA128983
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA128983 100 μL
1 options

Catalog No. PIPA128983

Supplier: Thermo Scientific PA128983

Rabbit Polyclonal Antibody

Campylobacter jejuni is a species of curved,long rod-shaped, Gram-negative microaerophilic, bacteria commonly found in animal feces. It is one of the most common causes of human gastroenteritis in the world. C. jejuni is commonly associated with poultry and naturally colonizes the G1 tract of many bird species.

PA1-28983 detects Campylobacter jejuni from bacteria samples.PA1-28983 has been successfully used in ELISA and immunocytochemistry applications.The PA1-28983 immunogen is: ATCC strain 29428
TRUSTED_SUSTAINABILITY

Specifications

Antigen Campylobacter jejuni
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 4mg/mL
Conjugate Unconjugated
Gene Alias C. jejuni
Host Species Rabbit
Immunogen ATCC strain 29428
Purification Method IgG fraction
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.