Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Campylobacter Rabbit anti-Bacteria, Biotin, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA173133

Catalog No. PA173133


Add to cart

Description

Description

The PA1-73133 antibody reacts with Campylobacter species. The antiserum has not been absorbed & may react with related microorganisms. PA1-73133 has been successfully used in immunofluorescence applications. The PA1-73133 antibody was raised against whole C. jejuni cells; ATCC strain 29428. Recommended short term storage, up to 6 months, is liquid at 4°C. For longer term storage, aliquot and freeze.

Specifications

Specifications

Campylobacter
Polyclonal
Biotin
Liquid
Whole C. jejuni cells; ATCC strain 29428.
Purified
RUO
Antibody
Polyclonal
Immunofluorescence
4-5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
Rabbit
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Bacteria
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit