Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CAMSAP1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP126644

Catalog No. NBP126644


Add to cart

Description

Description

CAMSAP1 Polyclonal antibody specifically detects CAMSAP1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

CAMSAP1
0.2 mg/mL
Western Blot 1:2000-1:10000, Immunoprecipitation 3-5 ug/mg lysate
NP_056262.3
bA100C15.1, bA100C15.1 (novel protein), calmodulin regulated spectrin-associated protein 1, calmodulin-regulated spectrin-associated protein 1, DKFZp434F195, DKFZp434G2311, FLJ31228, MGC163452, PRO2405
Rabbit
IgG
0.1 ml
Store at 4C. Do not freeze.
Polyclonal
Human
Immunoprecipitation, Western Blot
Unconjugated
TBS and 0.1% BSA with 0.09% Sodium Azide
Affinity Purified
CAMSAP1
The immunogen recognized by this antibody maps to a region between residue 1050 and 1100 of human calmodulin regulated spectrin-associated protein 1 using the numbering given in entry NP_056262.3 (GeneID 157922).
Immunogen affinity purified
RUO
Primary
157922
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only