Learn More
Candida albicans, Biotin, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA127145
Catalog No. PA127145
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Candida albicans is the most frequently isolated fungal pathogen of humans, affecting immuocompromised patients ranging from premature infants to AIDS sufferers. Systemic infections have an attributed mortality of 30-50%. C. albicans is a diploid organism which has eight sets of homologous chromosomes. Its genome size is about 16 Mb (haploid), about 30% greater than S. cerevisiae (baker's yeast).
PA1-27145 detects Candida albicans from fungi samples.PA1-27145 has been successfully used in immunohistochemistry. ELISA, and immunofluorescence procedures.The PA1-27145 immunogen is Candida albicans, type A.Store at 4ºC short term. For extended storage aliquot and store at -20ºC or below. Avoid freeze-thaw cycles.
Specifications
Specifications
Candida albicans
Polyclonal
Biotin
Purified
Rabbit
IgG
1mL
Store at -20°C
Primary
Fungi
ELISA, Immunocytochemistry, Immunodiffusion, Immunofluorescence, Immunohistochemistry
4mg/mL
PBS with 0.1% sodium azide; pH 7.2
C. albicans
Candida albicans, type A.
IgG fraction
RUO
Antibody
Polyclonal