Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Candida albicans Polyclonal Antibody, Biotin, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA173156
Description
The PA1-73156 antibody reacts with Candida albicans. PA1-73156 has been successfully used in Immunohistochemical, ELISA, Immunofluorescence and Western blot applications. The PA1-73156 antibody was raised against Candida albicans, type A.
Candida albicans Polyclonal antibody specifically detects Candida albicans in Yeast samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
Candida albicans | |
Polyclonal | |
Biotin | |
C. albicans | |
Candida albicans, type A. | |
1 mL | |
Primary | |
4° C | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
4-5 mg/mL | |
PBS with 0.1% sodium azide | |
Rabbit | |
purified | |
RUO | |
Yeast | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction